Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030138.2 | NP_084414.1 | 18 | 1965 | ||
GenBank mRNA | AB490692.1 | - | 19 | 1739 | ||
AK029343.1 | - | 18 | 1890 | |||
AK129041.1 | - | 18 | 1942 | |||
AK147319.1 | - | 17 | 1837 | |||
BC046455.1 | - | 5 | 508 |