Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178113.3 | NP_835214.2 | 17, 18 | 2500 | ||
GenBank mRNA | AK045277.1 | - | 4, 5 | 492 | ||
AK129046.1 | - | 16, 17 | 2026 | |||
AK139081.1 | - | 16, 17 | 2058 | |||
BC048190.1 | - | 17, 18 | 2500 | 6831876 | ||
DQ340802.1 | - | 16, 17 | 2155 |