Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178113.3 | NP_835214.2 | 4 | 832 | ||
GenBank mRNA | AK014555.1 | - | 3 | 621 | ||
AK019297.1 | - | 3 | 401 | |||
AK129046.1 | - | 3 | 358 | |||
AK139081.1 | - | 3 | 390 | |||
BC048190.1 | - | 4 | 832 | 6831876 | ||
DQ340802.1 | - | 3 | 487 |