Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Filip1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040397.4 | NP_001035487.2 | 5 | 1922 | ||
NM_001177871.1 | NP_001171342.1 | 2 | 1229 | |||
GenBank mRNA | AK019472.2 | - | 2 | 1230 | ||
AK132319.1 | - | 2 | 1230 | |||
BC062166.1 | - | 2 | 1212 | |||
BC098507.1 | - | 1 | 1066 | |||
EU623548.1 | - | 1 | 1041 | |||
EU704258.1 | - | 5 | 1769 |