Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030166.3 | NP_084442.1 | 1 | 435 | ||
GenBank mRNA | AB175682.1 | - | 1 | 45 | ||
AK019470.1 | - | 1 | 550 | |||
AK090330.1 | - | 1 | 129 | |||
AK163883.1 | - | 1 | 310 | |||
BC133711.1 | - | 1 | 435 | 40143107 |