Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhipl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019671.1 | - | 8, 9 | 1722 | ||
BC034362.1 | - | 7, 8 | 1467 | 3154701 |