Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enthd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183137.2 | NP_898960.2 | 2 | 135 | ||
GenBank mRNA | AK019105.1 | - | 2 | 106 | ||
AK154660.1 | - | 2 | 100 | |||
AK202725.1 | - | 2 | 50 | |||
AK214511.1 | - | 2 | 50 | |||
BB868415.1 | - | 2 | 136 | |||
BC049110.1 | - | 2 | 89 | |||
BC064313.1 | - | 2 | 89 | |||
BC094318.1 | - | 2 | 85 | |||
BC137580.1 | - | 2 | 104 | 8860599 | ||
BC144724.1 | - | 2 | 104 | 9053272 | ||
BY084429.1 | - | 2 | 105 |