Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114361.1 | NP_001107833.1 | 16 | 2059 | ||
NM_001114362.1 | NP_001107834.1 | 14 | 1852 | |||
NM_001286567.1 | NP_001273496.1 | 16 | 1962 | |||
NM_199466.3 | NP_955760.3 | 13 | 1723 | |||
GenBank mRNA | AK077527.1 | - | 8 | 887 | ||
AK144604.1 | - | 16 | 2060 | |||
BC032258.1 | - | 2 | 207 | |||
BC065178.1 | - | 16 | 1900 | 6827397 | ||
BC066099.1 | - | 13 | 1654 | 30533573 | ||
BC067011.1 | - | 13 | 1672 | 6405184 | ||
BC070427.1 | - | 14 | 1792 | 6413041 | ||
BC079619.1 | - | 13 | 1654 | 30533573 | ||
DQ230326.1 | - | 15 | 1742 |