Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081344.2 | NP_001074813.2 | 22 | 2999 | ||
GenBank mRNA | AF516607.1 | - | 18 | 2078 | ||
AK019788.1 | - | 5 | 740 | |||
AK029982.1 | - | 3 | 2750 | |||
AK122417.1 | - | 1 | 78 | |||
AK136998.1 | - | 1 | 1013 | |||
BC038042.1 | - | 2 | 367 | |||
BC094582.1 | - | 15 | 1824 |