Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198101.1 | NP_932769.1 | 14 | 1463 | ||
GenBank mRNA | AK019874.2 | - | 3 | 212 | ||
AK052221.1 | - | 14 | 1428 | |||
AK082337.1 | - | 14 | 1460 | |||
AK137177.1 | - | 12 | 1456 | |||
BC057037.1 | - | 14 | 1463 | 5702855 | ||
BC060628.1 | - | 14 | 1438 | 6847400 |