Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198101.1 | NP_932769.1 | 3 | 263 | ||
GenBank mRNA | AK043883.1 | - | 3 | 262 | ||
AK052221.1 | - | 3 | 230 | |||
AK082337.1 | - | 3 | 260 | |||
AK137177.1 | - | 1 | 256 | |||
AK172438.1 | - | 3 | 251 | |||
BC057037.1 | - | 3 | 263 | 5702855 | ||
BC060628.1 | - | 3 | 238 | 6847400 |