Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172436.3 | NP_766024.1 | 3, 4 | 236 | ||
GenBank mRNA | AK019150.1 | - | 3, 4 | 237 | ||
AK030595.1 | - | 3, 4 | 266 | |||
AK083156.1 | - | 3, 4 | 237 | |||
AK144958.1 | - | 3, 4 | 270 | |||
AK147948.1 | - | 3, 4 | 251 | |||
AK181457.1 | - | 2, 3 | 202 | |||
AK208043.1 | - | 3, 4 | 303 | |||
AK210791.1 | - | 2, 3 | 202 | |||
BC013845.1 | - | 3, 4 | 215 | |||
BC024952.1 | - | 3, 4 | 431 | |||
BC060505.1 | - | 3, 4 | 223 | 6311836 |