Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030205.4 | NP_084481.3 | 12 | 1112 | ||
GenBank mRNA | AK018739.1 | - | 12 | 1080 | ||
AK040408.1 | - | 1118 | ||||
AK088155.1 | - | 12 | 1080 | |||
AK143624.1 | - | 13 | 1235 | |||
AK150051.1 | - | 1135 | ||||
BC061006.1 | - | 12 | 1112 | 6536106 |