Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crispld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030209.3 | NP_084485.1 | 2 | 386 | ||
GenBank mRNA | AK019034.1 | - | 2 | 382 | ||
AK033858.1 | - | 2 | 347 | |||
AK132171.1 | - | 2 | 329 | |||
AK149909.1 | - | 2 | 343 | |||
AK149925.1 | - | 2 | 343 | |||
AK150180.1 | - | 2 | 343 | |||
AK154308.1 | - | 2 | 386 | |||
BC090642.1 | - | 2 | 380 | 30653388 | ||
CN527154.1 | - | 2 | 380 |