Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030215.3 | NP_084491.3 | 5 | 1740 | ||
GenBank mRNA | AB056151.1 | - | 5 | 1589 | ||
AF208046.1 | - | 5 | 985 | |||
AK041886.1 | - | 5 | 1696 | |||
AK050368.1 | - | 5 | 1742 | |||
AK082078.1 | - | 5 | 1655 | |||
AK167570.1 | - | 5 | 1741 | |||
AK170542.1 | - | 5 | 1733 | |||
AK170846.1 | - | 5 | 1721 | |||
AK209893.1 | - | 2 | 89 | |||
BC010482.1 | - | 5 | 1732 | 3492125 | ||
BC058744.1 | - | 5 | 1130 |