Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030225.4 | NP_084501.1 | 11 | 949 | ||
GenBank mRNA | AK019713.2 | - | 946 | |||
AK054053.1 | - | 11 | 903 | |||
AK149664.1 | - | 11 | 925 | |||
AK158877.1 | - | 11 | 950 | |||
AK168570.1 | - | 11 | 925 | |||
AK169943.1 | - | 11 | 903 | |||
AK197945.1 | - | 2 | 152 | |||
AK203473.1 | - | 2 | 152 | |||
AK205241.1 | - | 2 | 152 | |||
AK214016.1 | - | 2 | 206 | |||
AK220022.1 | - | 2 | 152 | |||
BC006702.1 | - | 11 | 886 | 3583936 | ||
BC024066.1 | - | 2 | 336 | |||
CT010197.1 | - | 11 | 885 |