Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190408.1 | NP_001177337.1 | 6 | 1797 | ||
NM_144560.3 | NP_653146.1 | 7 | 2055 | |||
GenBank mRNA | AF508323.1 | - | 4 | 2030 | ||
AF508324.1 | - | 5 | 1699 | |||
AK052229.1 | - | 7 | 2060 | |||
BC031785.1 | - | 6 | 1747 | 4194333 |