Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048189.4 | NP_001041654.2 | 2 | 293 | ||
NM_001284190.1 | NP_001271119.1 | 2 | 293 | |||
NM_030231.3 | NP_084507.1 | 2 | 293 | |||
GenBank mRNA | AK016502.1 | - | 2 | 258 | ||
AK019817.1 | - | 2 | 236 | |||
AK051109.1 | - | 2 | 236 | |||
BY277464.1 | - | 2 | 295 | |||
CJ049857.1 | - | 2 | 269 | |||
DQ867039.1 | - | 2 | 200 | |||
DQ867040.1 | - | 2 | 200 | |||
FN429928.1 | - | 2 | 119 |