Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170552.1 | NP_001164023.1 | 5 | 837 | ||
NM_030675.3 | NP_109600.2 | 5 | 791 | |||
GenBank mRNA | AF306509.1 | - | 5 | 504 | ||
AF310134.1 | - | 5 | 767 | |||
AK039264.1 | - | 812 | ||||
AK085107.1 | - | 5 | 819 | |||
AK144667.1 | - | 5 | 773 | |||
AK164306.1 | - | 2 | 3900 | |||
AK172215.1 | - | 3 | 4474 | |||
AY328895.1 | - | 4 | 473 | |||
AY464945.1 | - | 4 | 473 | |||
BC016087.1 | - | 5 | 741 | |||
BC044820.1 | - | 4 | 532 | |||
BC054819.1 | - | 5 | 882 | |||
BC085252.1 | - | 5 | 597 | |||
BC141042.1 | - | 5 | 635 | 9055654 |