Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170552.1 | NP_001164023.1 | 2 | 397 | ||
NM_030675.3 | NP_109600.2 | 2 | 351 | |||
GenBank mRNA | AF306509.1 | - | 2 | 64 | ||
AF310134.1 | - | 2 | 327 | |||
AK039264.1 | - | 372 | ||||
AK046128.1 | - | 1 | 2291 | |||
AK085107.1 | - | 2 | 379 | |||
AK144667.1 | - | 2 | 333 | |||
AK164306.1 | - | 1 | 1203 | |||
AK172215.1 | - | 1 | 3304 | |||
AY328895.1 | - | 1 | 33 | |||
AY464945.1 | - | 1 | 33 | |||
BB619038.1 | - | 2 | 333 | |||
BB853405.1 | - | 2 | 381 | |||
BB855141.1 | - | 2 | 351 | |||
BC016087.1 | - | 2 | 301 | |||
BC044820.1 | - | 1 | 92 | |||
BC054819.1 | - | 2 | 442 | |||
BC085252.1 | - | 2 | 157 | |||
BC141042.1 | - | 2 | 195 | 9055654 |