Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160415.1 | NP_001153887.1 | 8 | 1290 | ||
NM_030255.3 | NP_084531.2 | 7 | 1191 | |||
GenBank mRNA | AB461446.1 | - | 8 | 1181 | ||
AB461447.1 | - | 7 | 1082 | |||
AB461448.1 | - | 7 | 1082 | |||
AB461449.1 | - | 7 | 1082 | |||
AK044394.1 | - | 8 | 1250 | |||
AK049256.1 | - | 7 | 1151 | |||
AK049998.1 | - | 1145 | ||||
AK153719.1 | - | 7 | 1141 | |||
AK154035.1 | - | 1295 | ||||
BC003314.1 | - | 8 | 1272 | 3155422 | ||
BC007182.1 | - | 3 | 1427 | |||
EU707568.1 | - | 8 | 1181 | |||
EU707569.1 | - | 7 | 1013 | |||
EU707570.1 | - | 7 | 1082 | |||
EU707571.1 | - | 7 | 1013 | |||
GQ871501.1 | - | 8 | 1178 | |||
GQ871502.1 | - | 6 | 1022 | |||
GQ871503.1 | - | 7 | 1079 |