Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zxdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030260.3 | NP_084536.2 | 6 | 1825 | ||
NM_173002.3 | NP_766590.1 | 6 | 1825 | |||
GenBank mRNA | AK030287.1 | - | 6 | 1741 | ||
AK042190.1 | - | 6 | 1752 | |||
AK042392.1 | - | 6 | 1735 | |||
AK044429.1 | - | 6 | 1718 | |||
AK044529.1 | - | 6 | 1749 | |||
AK044948.1 | - | 6 | 1719 | |||
AK047079.1 | - | 6 | 1707 | |||
AK047538.1 | - | 6 | 1741 | |||
AK080307.1 | - | 6 | 1719 | |||
AK081005.1 | - | 6 | 1827 | |||
BC003332.1 | - | 6 | 1809 | 3256958 |