Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zxdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030260.3 | NP_084536.2 | 6 | 2203 | ||
NM_173002.3 | NP_766590.1 | 6 | 2203 | |||
GenBank mRNA | AK030287.1 | - | 6 | 2119 | ||
AK042190.1 | - | 6 | 2130 | |||
AK042392.1 | - | 6 | 2113 | |||
AK044429.1 | - | 6 | 2096 | |||
AK044529.1 | - | 6 | 2127 | |||
AK044948.1 | - | 6 | 2097 | |||
AK047079.1 | - | 6 | 2085 | |||
AK047538.1 | - | 6 | 2119 | |||
AK080307.1 | - | 6 | 2097 | |||
AK081005.1 | - | 6 | 2205 | |||
AK137347.1 | - | 2 | 357 | |||
BC003332.1 | - | 6 | 2185 | 3256958 |