Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030559.3 | NP_085036.3 | 10 | 1050 | ||
GenBank mRNA | AB056721.1 | - | 10 | 1008 | ||
AK049872.1 | - | 10 | 1007 | |||
AK162984.1 | - | 9 | 1157 | |||
AK169694.1 | - | 7 | 1587 | |||
BC025626.1 | - | 10 | 1016 | 5320572 |