Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030564.1 | NP_085041.1 | 3 | 374 | ||
GenBank mRNA | AF434815.1 | - | 2 | 269 | ||
AK045696.1 | - | 3 | 370 | |||
AK046274.1 | - | 3 | 356 | |||
AK046381.1 | - | 3 | 370 | |||
AK077221.1 | - | 3 | 371 | |||
AK132330.1 | - | 390 | ||||
AK137606.1 | - | 350 | ||||
AK150976.1 | - | 3 | 368 | |||
AK159753.1 | - | 3 | 356 | |||
AK166565.1 | - | 383 | ||||
BC004042.1 | - | 3 | 374 | 3496314 |