Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030697.2 | NP_109622.1 | 6 | 1683 | ||
GenBank mRNA | AK002623.1 | - | 6 | 1684 | ||
AK132011.1 | - | 6 | 1705 | |||
KJ650203.1 | - | 5 | 1614 | |||
KJ663713.1 | - | 5 | 1455 | |||
KJ663714.1 | - | 6 | 1476 |