Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271725.1 | NP_001258654.1 | 12 | 2403 | ||
NM_001271726.1 | NP_001258655.1 | 12 | 2439 | |||
NM_001271727.1 | NP_001258656.1 | 12 | 2708 | |||
NM_001271728.1 | NP_001258657.1 | 12 | 2506 | |||
NM_030706.3 | NP_109631.2 | 12 | 2502 | |||
GenBank mRNA | AB043550.1 | - | 12 | 2486 | ||
AF220017.1 | - | 12 | 2351 | |||
AK042752.1 | - | 4 | 495 | |||
AK045410.1 | - | 12 | 2499 | |||
AK079415.1 | - | 12 | 2653 | |||
AK141419.1 | - | 12 | 2349 | |||
AK147275.1 | - | 12 | 2616 | |||
AK147395.1 | - | 12 | 2497 | |||
AK164350.1 | - | 11 | 2205 | |||
AK220343.1 | - | 11 | 3945 | |||
BC058668.1 | - | 11 | 3314 | |||
BC058961.1 | - | 12 | 2506 | 6826143 |