Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290444.1 | NP_001277373.1 | 5 | 854 | ||
NM_030718.5 | NP_109643.3 | 6 | 989 | |||
GenBank mRNA | AB041039.1 | - | 5 | 877 | ||
AK033786.1 | - | 6 | 910 | |||
AK035261.1 | - | 6 | 974 | |||
BC116759.1 | - | 6 | 938 | 40126078 | ||
BC116761.1 | - | 6 | 938 | 40126080 |