Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030721.2 | NP_109646.2 | 9, 10 | 1062 | ||
GenBank mRNA | AJ278430.1 | - | 9, 10 | 1095 | ||
AK028818.1 | - | 9, 10 | 1483 | |||
AK053960.1 | - | 9, 10 | 1059 | |||
AK128922.1 | - | 9, 10 | 1022 | |||
AK154112.1 | - | 8, 9 | 1153 | |||
AK171598.1 | - | 1045 | ||||
BC044725.2 | - | 9, 10 | 1082 | 5374281 | ||
BC055019.1 | - | 9, 10 | 1062 | 4162223 |