Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030727.5 | NP_109652.3 | 5 | 571 | ||
GenBank mRNA | AF529192.1 | - | 5 | 571 | ||
AY024359.2 | - | 3 | 337 |