Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004140.2 | NP_001004140.2 | 8 | 1581 | ||
GenBank mRNA | AK034534.1 | - | 8 | 1536 | ||
AK132463.1 | - | 8 | 1569 | |||
AY692438.1 | - | 8 | 1581 | |||
BC145666.1 | - | 8 | 1574 | 40130969 | ||
BC146023.1 | - | 8 | 1574 | 40131326 |