Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024231.2 | NP_077193.1 | 3 | 162 | ||
GenBank mRNA | AK005244.1 | - | 3 | 174 | ||
BC002119.1 | - | 3 | 162 | 3491395 |