Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031251.4 | NP_112541.1 | 7 | 595 | ||
GenBank mRNA | AJ250670.1 | - | 6 | 493 | ||
AK054467.1 | - | 8 | 773 | |||
AK078511.1 | - | 7 | 607 | |||
AK079295.1 | - | 7 | 596 | |||
AK137143.1 | - | 7 | 638 | |||
AK170790.1 | - | 7 | 607 | |||
AK170828.1 | - | 7 | 618 | |||
BC005479.1 | - | 7 | 582 | 3484952 | ||
BC006033.1 | - | 7 | 582 |