Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031256.3 | NP_112546.1 | 2, 3 | 428 | ||
GenBank mRNA | AF286163.1 | - | 2, 3 | 217 | ||
BC012283.1 | - | 2, 3 | 168 | |||
BC031110.1 | - | 3, 4 | 236 | 4981486 | ||
BY730332.1 | - | 2, 3 | 428 |