Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031376.3 | NP_113553.1 | 2 | 519 | ||
GenBank mRNA | AF293806.1 | - | 2 | 519 | ||
AK089785.1 | - | 2 | 552 | |||
AK154326.1 | - | 2 | 547 | |||
AK170130.1 | - | 2 | 552 | |||
AK170925.1 | - | 2 | 552 | |||
AK171021.1 | - | 2 | 547 | |||
BC113141.1 | - | 2 | 478 | 40047744 |