Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sacm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030692.4 | NP_109617.1 | 10 | 806 | ||
GenBank mRNA | AJ245720.1 | - | 10 | 936 | ||
AJ297526.1 | - | 10 | 807 | |||
AK089174.1 | - | 10 | 890 | |||
AK122390.1 | - | 10 | 871 | |||
AK154759.1 | - | 10 | 906 | |||
AK161111.1 | - | 10 | 840 | |||
AK171080.1 | - | 10 | 890 | |||
BC005489.1 | - | 1 | 15 | |||
BC117752.1 | - | 10 | 807 | 40090556 | ||
BC117753.1 | - | 10 | 807 | 40090558 |