Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002238.2 | NP_001002238.1 | 16 | 2257 | ||
NM_001002240.2 | NP_001002240.1 | 16 | 2230 | |||
NM_001002241.2 | NP_001002241.1 | 15 | 2530 | |||
NM_031387.3 | NP_113564.2 | 16 | 2390 | |||
GenBank mRNA | AB067571.1 | - | 15 | 2211 | ||
AB183526.1 | - | 16 | 2257 | |||
AB183527.1 | - | 16 | 2230 | |||
AB183528.1 | - | 15 | 2530 | |||
AB183529.1 | - | 15 | 2094 | |||
AF285591.1 | - | 12 | 1584 | |||
AK029806.1 | - | 16 | 2391 | |||
BC129954.1 | - | 11 | 1516 | 40081321 | ||
BC129955.1 | - | 11 | 1516 | 40081323 |