Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crispld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031402.2 | NP_113579.2 | 9 | 1363 | ||
GenBank mRNA | AF329198.1 | - | 9 | 1360 | ||
AK132405.1 | - | 9 | 1101 | |||
AK133870.1 | - | 9 | 1364 | |||
AK134273.1 | - | 9 | 1299 | |||
AK137611.1 | - | 9 | 1364 | |||
BC040768.1 | - | 9 | 1692 | 4458200 |