Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031406.3 | NP_113583.2 | 7 | 1075 | ||
GenBank mRNA | AF314957.1 | - | 6 | 941 | ||
AK154118.1 | - | 7 | 1034 | |||
BC046982.1 | - | 7 | 995 | 6478046 |