Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031880.3 | NP_114086.3 | 10, 11 | 1591 | ||
GenBank mRNA | AF332512.1 | - | 8, 9 | 1411 | ||
AK033440.1 | - | 5, 6 | 962 | |||
AK076220.1 | - | 10, 11 | 1598 | |||
BC029623.1 | - | 9, 10 | 1578 | 4039780 | ||
BC055303.1 | - | 9, 10 | 1495 | 6439288 |