Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114386.1 | NP_001107858.1 | 24, 25 | 2653 | ||
NM_031881.2 | NP_114087.2 | 23, 24 | 2702 | |||
GenBank mRNA | AF277232.1 | - | 23, 24 | 2335 | ||
AK004969.1 | - | 23, 24 | 2395 | |||
AK031842.1 | - | 22, 23 | 2318 | |||
AK042621.1 | - | 23, 24 | 2276 | |||
AK163497.1 | - | 21, 22 | 2123 | |||
AK197029.1 | - | 2, 3 | 179 | |||
AK199884.1 | - | 3, 4 | 200 | |||
AK202076.1 | - | 3, 4 | 200 | |||
AK211578.1 | - | 3, 4 | 220 | |||
BC039746.1 | - | 23, 24 | 2346 | 3157137 | ||
BC071210.1 | - | 23, 24 | 2323 | 4972426 | ||
BC082281.1 | - | 20, 21 | 2060 |