Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081216.1 | NP_001074685.1 | 7 | 705 | ||
GenBank mRNA | AB049460.1 | - | 7 | 624 | ||
AJ303103.1 | - | 7 | 601 | |||
AK032797.1 | - | 7 | 735 | |||
AK042284.1 | - | 4 | 871 | |||
AK044935.1 | - | 7 | 632 | |||
AK049736.1 | - | 7 | 640 | |||
AK050025.1 | - | 7 | 650 | |||
AK136530.1 | - | 7 | 643 | |||
BC022102.1 | - | 7 | 646 | |||
BC037036.1 | - | 7 | 634 | |||
BC039765.1 | - | 7 | 657 | |||
BC049950.1 | - | 7 | 635 |