Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081216.1 | NP_001074685.1 | 35 | 4233 | ||
GenBank mRNA | AF310251.1 | - | 12 | 1497 | ||
AJ303103.1 | - | 35 | 4129 | |||
AK162189.1 | - | 16 | 1803 | |||
BC037950.1 | - | 9 | 866 |