Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252580.1 | NP_001239509.1 | 10 | 1326 | ||
NM_019729.3 | NP_062703.2 | 10 | 1293 | |||
GenBank mRNA | AB045709.1 | - | 9 | 1067 | ||
AF057146.1 | - | 9 | 1084 | |||
AK122195.1 | - | 10 | 1204 | |||
AK129003.1 | - | 10 | 1294 | |||
AK134058.1 | - | 10 | 1248 | |||
AK142881.1 | - | 7 | 791 | |||
AK153015.1 | - | 9 | 1041 | |||
AK163469.1 | - | 10 | 1226 | |||
BC038123.1 | - | 10 | 1278 | |||
BC050947.1 | - | 10 | 1258 | 6825981 | ||
BC061465.1 | - | 10 | 1258 | 6825981 | ||
BC066126.1 | - | 10 | 1256 | 30301665 |