Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252580.1 | NP_001239509.1 | 8 | 1043 | ||
NM_019729.3 | NP_062703.2 | 8 | 1010 | |||
GenBank mRNA | AB045709.1 | - | 7 | 784 | ||
AF057146.1 | - | 7 | 801 | |||
AK122195.1 | - | 8 | 921 | |||
AK129003.1 | - | 8 | 1011 | |||
AK134058.1 | - | 8 | 965 | |||
AK142881.1 | - | 5 | 508 | |||
AK153015.1 | - | 7 | 758 | |||
AK163469.1 | - | 8 | 943 | |||
BC038123.1 | - | 8 | 995 | |||
BC050947.1 | - | 8 | 975 | 6825981 | ||
BC061465.1 | - | 8 | 975 | 6825981 | ||
BC066126.1 | - | 8 | 973 | 30301665 |