Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam126a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053090.2 | NP_444320.2 | 5 | 592 | ||
GenBank mRNA | AK155209.1 | - | 5 | 593 | ||
AK161265.1 | - | 5 | 786 | |||
BC056381.1 | - | 5 | 531 | 6852011 | ||
BC060692.1 | - | 5 | 579 | 6410470 |