Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam126a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053090.2 | NP_444320.2 | 11 | 1569 | ||
GenBank mRNA | BC056381.1 | - | 11 | 1508 | 6852011 | |
BC060692.1 | - | 11 | 1556 | 6410470 |