Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170395.1 | NP_001163866.1 | 4 | 608 | ||
NM_053094.2 | NP_444324.2 | 4 | 608 | |||
GenBank mRNA | AF274883.1 | - | 4 | 603 | ||
BC144848.1 | - | 4 | 559 | 9053396 | ||
BC145793.1 | - | 4 | 559 | 40131096 | ||
DQ058616.1 | - | 4 | 564 | |||
DQ058617.1 | - | 4 | 564 |