Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053100.2 | NP_444330.2 | 5 | 1786 | ||
GenBank mRNA | AF220035.1 | - | 4 | 405 | ||
AF281047.1 | - | 5 | 1135 | |||
AK052763.1 | - | 5 | 521 | |||
AK079847.1 | - | 3 | 225 | |||
AK155303.1 | - | 5 | 1733 | |||
BC029283.1 | - | 5 | 1720 | |||
BC037065.1 | - | 5 | 1071 | |||
BC060094.1 | - | 5 | 1016 | |||
BC082298.1 | - | 5 | 1661 | |||
BC152343.1 | - | 5 | 1122 | 40093266 | ||
BC152344.1 | - | 5 | 1122 | 40093269 |