Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053100.2 | NP_444330.2 | 1 | 966 | ||
GenBank mRNA | AF281047.1 | - | 1 | 315 | ||
AK155303.1 | - | 1 | 913 | |||
BC029283.1 | - | 1 | 900 | |||
BC037065.1 | - | 1 | 251 | |||
BC060094.1 | - | 1 | 195 | |||
BC082298.1 | - | 1 | 841 | |||
BC152343.1 | - | 1 | 302 | 40093266 | ||
BC152344.1 | - | 1 | 302 | 40093269 |
Gene Symbol: Gm9798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC024694.1 | - | 1 | 166 |